missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H,1H,5H-Octafluoropentyl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™

Manufacturer:  TCI America O048125G

 View more versions of this product

Catalog No. O048125G



Specifications

Specifications

1H,1H,5H-Octafluoropentyl Methacrylate (stabilized with MEHQ)
Colorless
355-93-1
MFCD00039278
ZNJXRXXJPIFFAO-UHFFFAOYSA-N
2,2,3,3,4,4,5,5-octafluoropentyl 2-methylprop-2-enoate
67739
Clear Liquid at 20°C
88°C
25g
C9H8F8O2
1h,1h,5h-octafluoropentyl methacrylate, 2,2,3,3,4,4,5,5-octafluoropentyl methacrylate, octafluoropentyl methacrylate, 1h,1h,5h-perfluoropentyl methacrylate, 1h,1h,5h-octafluoropentyl methacrylate stabilized with mehq, 2-propenoic acid, 2-methyl-, 2,2,3,3,4,4,5,5-octafluoropentyl ester, methacrylic acid 1h,1h,5h-perfluoropentyl ester, methacrylic acid 1h,1h,5h-octafluoropentyl ester, octafluoropentyl methacrylate polymer, 1h,1h,5h-octafluoropentylmethacrylate
CC(=C)C(=O)OCC(C(C(C(F)F)(F)F)(F)F)(F)F
300.148
300.15
≥98.0% (GC)

Chemical Identifiers

355-93-1
300.148
ZNJXRXXJPIFFAO-UHFFFAOYSA-N
67739
CC(=C)C(=O)OCC(C(C(C(F)F)(F)F)(F)F)(F)F
C9H8F8O2
MFCD00039278
1h,1h,5h-octafluoropentyl methacrylate, 2,2,3,3,4,4,5,5-octafluoropentyl methacrylate, octafluoropentyl methacrylate, 1h,1h,5h-perfluoropentyl methacrylate, 1h,1h,5h-octafluoropentyl methacrylate stabilized with mehq, 2-propenoic acid, 2-methyl-, 2,2,3,3,4,4,5,5-octafluoropentyl ester, methacrylic acid 1h,1h,5h-perfluoropentyl ester, methacrylic acid 1h,1h,5h-octafluoropentyl ester, octafluoropentyl methacrylate polymer, 1h,1h,5h-octafluoropentylmethacrylate
2,2,3,3,4,4,5,5-octafluoropentyl 2-methylprop-2-enoate
Safety and Handling

Safety and Handling

TSCA : No

Documents