Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H,1H,2H-Perfluoro-1-decene, 99%

$51.38 - $473.12

Chemical Identifiers

Chemical Name or Material 1H,1H,2H-Perfluoro-1-decene
CAS 21652-58-4
Molecular Formula C10H3F17
Linear Formula CF3(CF2)7CH=CH2
Formula Weight 446.11
MDL Number MFCD00039246
Beilstein 2066558
Synonym 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluoro-1-decene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2023106

  SDS
alfa aesar™
B2023106
5g Each for $51.38

AAB2023114

  SDS
alfa aesar™
B2023114
25g Each for $165.74

AAB2023122

  SDS
alfa aesar™
B2023122
100g Each for $473.12
Description & Specifications

Specifications

Formula Weight 446.11
Chemical Name or Material 1H,1H,2H-Perfluoro-1-decene
TSCA TSCA
Assay 99%
Molecular Formula C10H3F17
Linear Formula CF3(CF2)7CH=CH2
CAS 21652-58-4
Beilstein 2066558
Synonym 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluoro-1-decene
Health Hazard 1 Warning
Quantity 5g
MDL Number MFCD00039246
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 125°C (257°F)
Boiling Point 146° to 147°C

Applications
1H,1H,2H-Perfluoro-1-decene can be used to prepare perfluorononan-1-oic acid. Appliance Paint, Boat Paint, Buidling. It con be used as monomer and key intermediate of fluoro silicon surfactant.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.