missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H,1H,2H-Heptadecafluoro-1-decene, TCI America™

$27.50 - $80.97

Chemical Identifiers

CAS 21652-58-4
Molecular Formula C10H3F17
Molecular Weight (g/mol) 446.107
MDL Number MFCD00039246
InChI Key NKAMGQZDVMQEJL-UHFFFAOYSA-N
Synonym 1h,1h,2h-perfluoro-1-decene, perfluoro-n-octyl ethylene, perfluorooctyl ethylene, tridecafluoro 1-decene, 1h,1h,2h-perfluorodec-1-ene, 1h,1h,2h-heptadecafluoro-1-decene, perfluoroctyl ethylene, cf3 cf2 7ch=ch2, 1-heptadecafluorooctyl ethene, 1h,1h,2h-perfluoro-1-decene
PubChem CID 88990
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene
SMILES C=CC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H084625G View Documents TCI America
H084625G
25g Each for $80.97
 
H08465G View Documents TCI America
H08465G
5g Each for $27.50
 
Specifications

Chemical Identifiers

21652-58-4
446.107
NKAMGQZDVMQEJL-UHFFFAOYSA-N
88990
C=CC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
C10H3F17
MFCD00039246
1h,1h,2h-perfluoro-1-decene, perfluoro-n-octyl ethylene, perfluorooctyl ethylene, tridecafluoro 1-decene, 1h,1h,2h-perfluorodec-1-ene, 1h,1h,2h-heptadecafluoro-1-decene, perfluoroctyl ethylene, cf3 cf2 7ch=ch2, 1-heptadecafluorooctyl ethene, 1h,1h,2h-perfluoro-1-decene
3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene

Specifications

147°C
25g
C10H3F17
1993
NKAMGQZDVMQEJL-UHFFFAOYSA-N
3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene
88990
Clear Liquid at 20°C
1H,1H,2H-Heptadecafluoro-1-decene
Yellow
21652-58-4
MFCD00039246
1h,1h,2h-perfluoro-1-decene, perfluoro-n-octyl ethylene, perfluorooctyl ethylene, tridecafluoro 1-decene, 1h,1h,2h-perfluorodec-1-ene, 1h,1h,2h-heptadecafluoro-1-decene, perfluoroctyl ethylene, cf3 cf2 7ch=ch2, 1-heptadecafluorooctyl ethene, 1h,1h,2h-perfluoro-1-decene
C=CC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
446.107
446.11
≥97.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3594

TSCA : Yes