Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H,1H,2H,3H,3H-Perfluorononane-1,2-diol, 95%

$105.47 - $406.82

Chemical Identifiers

Chemical Name or Material 1H,1H,2H,3H,3H-Perfluorononane-1,2-diol
CAS 107650-06-6
Molecular Formula C9H7F13O2
Formula Weight 394.13
MDL Number MFCD00042262
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1659806

  SDS
alfa aesar™
L1659806
5g Each for $105.47

AAL1659814

  SDS
alfa aesar™
L1659814
25g Each for $406.82
Description & Specifications

Specifications

Formula Weight 394.13
Chemical Name or Material 1H,1H,2H,3H,3H-Perfluorononane-1,2-diol
Assay 95%
CAS 107650-06-6
Molecular Formula C9H7F13O2
MDL Number MFCD00042262
Boiling Point 120° to 130°C
Quantity 5g
Melting Point 60° to 64°C
Flash Point >150°C (302°F)