missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H,1H,2H,2H-Tridecafluoro-1-n-octanol, TCI America™

Manufacturer:  TCI America T252825G

 View more versions of this product

Catalog No. T252825G



Specifications

Specifications

1H,1H,2H,2H-Tridecafluoro-1-n-octanol
Yellow
647-42-7
MFCD00042143
GRJRKPMIRMSBNK-UHFFFAOYSA-N
3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctan-1-ol
69537
Clear Liquid at 20°C
87°C
25g
C8H5F13O
3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-octanol, 2-perfluorohexyl ethanol, 1h,1h,2h,2h-perfluorooctan-1-ol, 1h,1h,2h,2h-perfluorooctanol, 1h,1h,2h,2h-perfluoro-1-octanol, unii-g2r5yo5n3v, 1-octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro, 1h,1h,2h,2h-tridecafluoro-1-n-octanol, g2r5yo5n3v, 1,1,2,2-tetrahydroperfluoro-1-octanol
C(CO)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
364.106
364.11
≥98.0% (GC)

Chemical Identifiers

647-42-7
364.106
GRJRKPMIRMSBNK-UHFFFAOYSA-N
69537
C(CO)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
C8H5F13O
MFCD00042143
3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-octanol, 2-perfluorohexyl ethanol, 1h,1h,2h,2h-perfluorooctan-1-ol, 1h,1h,2h,2h-perfluorooctanol, 1h,1h,2h,2h-perfluoro-1-octanol, unii-g2r5yo5n3v, 1-octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro, 1h,1h,2h,2h-tridecafluoro-1-n-octanol, g2r5yo5n3v, 1,1,2,2-tetrahydroperfluoro-1-octanol
3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctan-1-ol
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2402

TSCA : Yes

Documents