Learn More
1H,1H,2H,2H-Perfluorooctylmethyldichlorosilane, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1660506
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
189°C to 190°C | |
73609-36-6 | |
C9H7Cl2F13Si | |
UN2920 | |
51°C (123°F) | |
VBDMVWQNRXVEGC-UHFFFAOYSA-N | |
dichloro-methyl-(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane | |
123573 | |
97% | |
Moisture sensitive |
1H,1H,2H,2H-Perfluorooctylmethyldichlorosilane | |
5 g | |
MFCD00042360 | |
6153667 | |
1h,1h,2h,2h-perfluorooctyl methyldichlorosilane, 1h,1h,2h,2h-perfluorooctylmethyldichlorosilane, dichloro methyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, dichloromethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, tridecafluoro-1,1,2,2-tetrahydrooctyl methyldichlorosilane, tridecafluoro-1,1,2,2-tetrahydrooctyl-1-methyldichlorosilane, dichloro-methyl-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, bis chloranyl-methyl-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecakis fluoranyl octyl silane, silane, dichloromethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl, 2-tridecafluorohexyl ethyldichloro methyl silane | |
C[Si](Cl)(Cl)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F | |
461.12 | |
461.13 | |
1.35 | |
1.55 |
Chemical Identifiers
73609-36-6 | |
461.12 | |
VBDMVWQNRXVEGC-UHFFFAOYSA-N | |
123573 | |
C[Si](Cl)(Cl)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
C9H7Cl2F13Si | |
MFCD00042360 | |
1h,1h,2h,2h-perfluorooctyl methyldichlorosilane, 1h,1h,2h,2h-perfluorooctylmethyldichlorosilane, dichloro methyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, dichloromethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, tridecafluoro-1,1,2,2-tetrahydrooctyl methyldichlorosilane, tridecafluoro-1,1,2,2-tetrahydrooctyl-1-methyldichlorosilane, dichloro-methyl-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, bis chloranyl-methyl-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecakis fluoranyl octyl silane, silane, dichloromethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl, 2-tridecafluorohexyl ethyldichloro methyl silane | |
dichloro-methyl-(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber : 277-551-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only