missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane, 97%

Manufacturer:  Alfa Aesar™ 4454309

 View more versions of this product

Catalog No. AA4454309



Specifications

Specifications

1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane
C10H10ClF13Si
UN2986
1h,1h,2h,2h-perfluorooctyldimethylchlorosilane, chlorodimethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, tridecafluoro-1,1,2,2-tetrahydrooctyl dimethylchlorosilane, 2-tridecafluorohexyl ethyldimethylchlorosilane, tridecafluoro-1,1,2,2-tetrahydrooctyl-1-dimethylchlorosilane, tridecafluoro-1,1,2,2-tetrahydrooctyl-1-dimethylchlorosilane, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyldimethylchlorosilane, chloro-dimethyl-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, chlorodimethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-n-octyl silane, silane, chlorodimethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl
C[Si](C)(CCC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)Cl
440.704
440.71
97%
1.3453
189°C to 191°C
10g
102488-47-1
MFCD00042359
7040835
AKYGPHVLITVSJE-UHFFFAOYSA-N
chloro-dimethyl-(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
145382
Liquid
1.461g/mL
Moisture Sensitive
Colorless
52°C (180°F)

Chemical Identifiers

102488-47-1
440.704
AKYGPHVLITVSJE-UHFFFAOYSA-N
145382
C[Si](C)(CCC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)Cl
C10H10ClF13Si
MFCD00042359
1h,1h,2h,2h-perfluorooctyldimethylchlorosilane, chlorodimethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, tridecafluoro-1,1,2,2-tetrahydrooctyl dimethylchlorosilane, 2-tridecafluorohexyl ethyldimethylchlorosilane, tridecafluoro-1,1,2,2-tetrahydrooctyl-1-dimethylchlorosilane, tridecafluoro-1,1,2,2-tetrahydrooctyl-1-dimethylchlorosilane, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyldimethylchlorosilane, chloro-dimethyl-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, chlorodimethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-n-octyl silane, silane, chlorodimethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl
chloro-dimethyl-(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, FLAMMABLE, N.O.S.

SDS Documents