missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H,1H,2H,2H-Perfluorodecylmethyldichlorosilane, 95%

$49.04 - $499.65

Chemical Identifiers

CAS 3102-79-2
Molecular Formula C11H7Cl2F17Si
Molecular Weight (g/mol) 561.135
MDL Number MFCD00042342
InChI Key PVBMWIXRKLGXPI-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorodecylmethyldichlorosilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl methyldichlorosilane, dichloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl methylsilane, dichloro-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl-methylsilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl-1-methyldichlorosilane, 1h,1h,2h,2h-perfluorodecyldichloromethylsilane, bis chloranyl-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecakis fluoranyl decyl-methyl-silane
PubChem CID 137826
IUPAC Name dichloro-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)-methylsilane
SMILES C[Si](CCC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(Cl)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1658306 View Documents Alfa Aesar™
L1658306
5g Each for $139.70
 
AAL1658303 View Documents Alfa Aesar™
L1658303
1g Each for $49.04
 
AAL1658314 View Documents Alfa Aesar™
L1658314
25g Each for $499.65
 
Specifications

Chemical Identifiers

3102-79-2
561.135
PVBMWIXRKLGXPI-UHFFFAOYSA-N
137826
C[Si](CCC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(Cl)Cl
C11H7Cl2F17Si
MFCD00042342
1h,1h,2h,2h-perfluorodecylmethyldichlorosilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl methyldichlorosilane, dichloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl methylsilane, dichloro-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl-methylsilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl-1-methyldichlorosilane, 1h,1h,2h,2h-perfluorodecyldichloromethylsilane, bis chloranyl-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecakis fluoranyl decyl-methyl-silane
dichloro-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)-methylsilane

Specifications

3102-79-2
MFCD00042342
5889048
PVBMWIXRKLGXPI-UHFFFAOYSA-N
dichloro-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)-methylsilane
137826
95%
5g
C11H7Cl2F17Si
UN2987
1h,1h,2h,2h-perfluorodecylmethyldichlorosilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl methyldichlorosilane, dichloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl methylsilane, dichloro-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl-methylsilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl-1-methyldichlorosilane, 1h,1h,2h,2h-perfluorodecyldichloromethylsilane, bis chloranyl-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecakis fluoranyl decyl-methyl-silane
C[Si](CCC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(Cl)Cl
561.135
561.14
205°C to 207°C
1H,1H,2H,2H-Perfluorodecylmethyldichlorosilane
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.