Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H,1H,2H,2H-Perfluoro-1-iodododecane, 97%

$89.80 - $333.00

Chemical Identifiers

Chemical Name or Material 1H,1H,2H,2H-Perfluoro-1-iodododecane
CAS 2043-54-1
Molecular Formula CF3(CF2)9CH2CH2I
Formula Weight 674.04
MDL Number MFCD00039412
Beilstein 1897869
Synonym 1-Iodo-1H,1H,2H,2H-perfluorododecane; 1H,1H,2H,2H-Perfluorododecyl iodide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1652706

  SDS
alfa aesar™
L1652706
5g Each for $89.80

AAL1652714

  SDS
alfa aesar™
L1652714
25g Each for $333.00
Description & Specifications

Specifications

Formula Weight 674.04
Chemical Name or Material 1H,1H,2H,2H-Perfluoro-1-iodododecane
MDL Number MFCD00039412
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Molecular Formula CF3(CF2)9CH2CH2I
CAS 2043-54-1
Assay 97%
TSCA TSCA
Melting Point 85° to 87°C
Synonym 1-Iodo-1H,1H,2H,2H-perfluorododecane; 1H,1H,2H,2H-Perfluorododecyl iodide
Quantity 5g
Odor Odorless
Boiling Point 108°C (10mmHg)
Beilstein 1897869
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.