Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H,1H,2H,2H-Perfluoro-1-iododecane, 97%

$45.86 - $152.39

Chemical Identifiers

Chemical Name or Material 1H,1H,2H,2H-Perfluoro-1-iododecane
CAS 2043-53-0
Molecular Formula C10F17H4I
Linear Formula CF3(CF2)7CH2CH2I
Formula Weight 574.02
MDL Number MFCD00039411
Beilstein 1894667
Synonym 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluoro-10-iododecane; 1-Iodo-1H,1H,2H,2H-perfluorodecane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2089414

 
alfa aesar™
B2089414
25g Each for $152.39

AAB2089406

 
alfa aesar™
B2089406
5g Each for $45.86
Description & Specifications

Specifications

Formula Weight 574.02
Chemical Name or Material 1H,1H,2H,2H-Perfluoro-1-iododecane
TSCA TSCA
Assay 97%
Molecular Formula C10F17H4I
Linear Formula CF3(CF2)7CH2CH2I
MDL Number MFCD00039411
CAS 2043-53-0
Beilstein 1894667
Synonym 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluoro-10-iododecane; 1-Iodo-1H,1H,2H,2H-perfluorodecane
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Boiling Point 226°C
Melting Point 53° to 57°C
Quantity 25g

Applications
It is used in that preparation of a chemical compounds employed in high-performance organic thin-film transistors. It also finds it application as an appliance in boat and building paint.

Solubility
Insoluble in water.

Notes
Light sensitive. Store away from alkali metals bases and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.