Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H,1H,2H,2H-Perfluoro-1-dodecanol, 96%


Quantity 1g

Manufacturer: alfa aesar™  L1658903

 View more versions of this product

Catalog No. AAL1658903

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 564.14
Assay 96%
Molecular Formula C12H5F21O
MDL Number MFCD00039545
CAS 865-86-1
Beilstein 2029867
Melting Point 90° to 95°C
Quantity 1g
Chemical Name or Material 1H,1H,2H,2H-Perfluoro-1-dodecanol
TSCA TSCA
Linear Formula CF3(CF2)9CH2CH2OH
Boiling Point 145°C (10mmHg)

Specifications

Chemical Name or Material 1H,1H,2H,2H-Perfluoro-1-dodecanol
CAS 865-86-1
Molecular Formula C12H5F21O
Linear Formula CF3(CF2)9CH2CH2OH
Formula Weight 564.14
MDL Number MFCD00039545
Beilstein 2029867

Applications
1H,1H,2H,2H-Perfluoro-1-dodecanol is used in fabric processing, coating, appliance paint and boat paint.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids and oxidizing agents.