missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H,1H,2H,2H-Nonafluorohexyl Methacrylate (stabilized with MEHQ), TCI America™

$55.00 - $161.95

Chemical Identifiers

CAS 1799-84-4
Molecular Formula C10H9F9O2
Molecular Weight (g/mol) 332.166
MDL Number MFCD00236094
InChI Key TYNRPOFACABVSI-UHFFFAOYSA-N
Synonym Methacrylic Acid 1H,1H,2H,2H-Nonafluorohexyl Ester, 3,3,4,4,5,5,6,6,6-Nonafluorohexyl Methacrylate, Methacrylic Acid 3,3,4,4,5,5,6,6,6-Nonafluorohexyl Ester
PubChem CID 74534
IUPAC Name 3,3,4,4,5,5,6,6,6-nonafluorohexyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCC(C(C(C(F)(F)F)(F)F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N10145G View Documents TCI America
N10145G
5g Each for $55.00
 
N101425G View Documents TCI America
N101425G
25g Each for $161.95
 
Specifications

Chemical Identifiers

1799-84-4
332.166
TYNRPOFACABVSI-UHFFFAOYSA-N
74534
CC(=C)C(=O)OCCC(C(C(C(F)(F)F)(F)F)(F)F)(F)F
C10H9F9O2
MFCD00236094
Methacrylic Acid 1H,1H,2H,2H-Nonafluorohexyl Ester, 3,3,4,4,5,5,6,6,6-Nonafluorohexyl Methacrylate, Methacrylic Acid 3,3,4,4,5,5,6,6,6-Nonafluorohexyl Ester
3,3,4,4,5,5,6,6,6-nonafluorohexyl 2-methylprop-2-enoate

Specifications

Colorless
1799-84-4
MFCD00236094
TYNRPOFACABVSI-UHFFFAOYSA-N
3,3,4,4,5,5,6,6,6-nonafluorohexyl 2-methylprop-2-enoate
74534
Clear Liquid at 20°C
1H,1H,2H,2H-Nonafluorohexyl Methacrylate (stabilized with MEHQ)
5g
C10H9F9O2
Methacrylic Acid 1H,1H,2H,2H-Nonafluorohexyl Ester, 3,3,4,4,5,5,6,6,6-Nonafluorohexyl Methacrylate, Methacrylic Acid 3,3,4,4,5,5,6,6,6-Nonafluorohexyl Ester
CC(=C)C(=O)OCCC(C(C(C(F)(F)F)(F)F)(F)F)(F)F
332.166
332.17
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : Yes