missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1H,1H,2H,2H-Nonafluorohexyl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™
Supplier: TCI America N10145G
Specifications
1H,1H,2H,2H-Nonafluorohexyl Methacrylate (stabilized with MEHQ) | |
Colorless | |
MFCD00236094 | |
Methacrylic Acid 1H,1H,2H,2H-Nonafluorohexyl Ester, 3,3,4,4,5,5,6,6,6-Nonafluorohexyl Methacrylate, Methacrylic Acid 3,3,4,4,5,5,6,6,6-Nonafluorohexyl Ester | |
CC(=C)C(=O)OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F | |
332.17 | |
332.17 | |
Liquid |
1799-84-4 | |
C10H9F9O2 | |
5 g | |
TYNRPOFACABVSI-UHFFFAOYSA-N | |
3,3,4,4,5,5,6,6,6-nonafluorohexyl 2-methylprop-2-enoate | |
74534 | |
≥98.0% (GC) |
Chemical Identifiers
1799-84-4 | |
332.17 | |
TYNRPOFACABVSI-UHFFFAOYSA-N | |
74534 | |
CC(=C)C(=O)OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
C10H9F9O2 | |
MFCD00236094 | |
Methacrylic Acid 1H,1H,2H,2H-Nonafluorohexyl Ester, 3,3,4,4,5,5,6,6,6-Nonafluorohexyl Methacrylate, Methacrylic Acid 3,3,4,4,5,5,6,6,6-Nonafluorohexyl Ester | |
3,3,4,4,5,5,6,6,6-nonafluorohexyl 2-methylprop-2-enoate |
Safety and Handling
TSCA : Yes