missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H,1H,2H,2H-Nonafluoro-1-hexanol, TCI America™

$50.42 - $157.36

Chemical Identifiers

CAS 2043-47-2
Molecular Formula C6H5F9O
Molecular Weight (g/mol) 264.091
MDL Number MFCD00039543
InChI Key JCMNMOBHVPONLD-UHFFFAOYSA-N
Synonym 2-(Perfluorobutyl)ethanol
PubChem CID 74883
IUPAC Name 3,3,4,4,5,5,6,6,6-nonafluorohexan-1-ol
SMILES C(CO)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N06005G View Documents TCI America
N06005G
5g Each for $50.42
 
N060025G View Documents TCI America
N060025G
25g Each for $157.36
 
Specifications

Chemical Identifiers

2043-47-2
264.091
JCMNMOBHVPONLD-UHFFFAOYSA-N
74883
C(CO)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F
C6H5F9O
MFCD00039543
2-(Perfluorobutyl)ethanol
3,3,4,4,5,5,6,6,6-nonafluorohexan-1-ol

Specifications

Colorless
2043-47-2
MFCD00039543
JCMNMOBHVPONLD-UHFFFAOYSA-N
3,3,4,4,5,5,6,6,6-nonafluorohexan-1-ol
74883
Clear Liquid at 20°C
1H,1H,2H,2H-Nonafluoro-1-hexanol
5g
C6H5F9O
2-(Perfluorobutyl)ethanol
C(CO)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F
264.091
264.09
≥97.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2402

TSCA : Yes