missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H,1H,2H,2H-Heptadecafluorodecyl Iodide 98.0+%, TCI America™

$41.14 - $120.16

Chemical Identifiers

CAS 2043-53-0
Molecular Formula C10H4F17I
Molecular Weight (g/mol) 574.019
MDL Number MFCD00039411
InChI Key XVKJSLBVVRCOIT-UHFFFAOYSA-N
Synonym 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-10-iodo-decane, 1-iodo-1h,1h,2h,2h-perfluorodecane, 1h,1h,2h,2h-heptadecafluorodecyl iodide, 1h,1h,2h,2h-perfluoro-1-iododecane, 1h,1h,2h,2h-perfluorodecyl iodide, 1h,1h,2h,2h-perfluorodecyliodide, 2-perfluorooctyl ethyl iodide, acmc-20ap95, c8f17ch2ch2i, decane, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-10-iodo
PubChem CID 74885
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-10-iododecane
SMILES C(CI)C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H10845G View Documents Tci America
H10845G
5g Each for $41.14
 
H108425G View Documents Tci America
H108425G
25g Each for $120.16
 
Specifications

Chemical Identifiers

2043-53-0
574.019
XVKJSLBVVRCOIT-UHFFFAOYSA-N
74885
C(CI)C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
C10H4F17I
MFCD00039411
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-10-iodo-decane, 1-iodo-1h,1h,2h,2h-perfluorodecane, 1h,1h,2h,2h-heptadecafluorodecyl iodide, 1h,1h,2h,2h-perfluoro-1-iododecane, 1h,1h,2h,2h-perfluorodecyl iodide, 1h,1h,2h,2h-perfluorodecyliodide, 2-perfluorooctyl ethyl iodide, acmc-20ap95, c8f17ch2ch2i, decane, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-10-iodo
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-10-iododecane

Specifications

96°C
58°C
2043-53-0
MFCD00039411
XVKJSLBVVRCOIT-UHFFFAOYSA-N
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-10-iododecane
74885
Crystal-Powder at 20°C
1H,1H,2H,2H-Heptadecafluorodecyl Iodide
White-Yellow
5g
C10H4F17I
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-10-iodo-decane, 1-iodo-1h,1h,2h,2h-perfluorodecane, 1h,1h,2h,2h-heptadecafluorodecyl iodide, 1h,1h,2h,2h-perfluoro-1-iododecane, 1h,1h,2h,2h-perfluorodecyl iodide, 1h,1h,2h,2h-perfluorodecyliodide, 2-perfluorooctyl ethyl iodide, acmc-20ap95, c8f17ch2ch2i, decane, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-10-iodo
C(CI)C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
574.019
574.02
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-4011

TSCA : Yes