Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H,1H,11H-Perfluoroundecyl methacrylate, 95%

$62.96 - $224.73

Chemical Identifiers

Chemical Name or Material 1H,1H,11H-Perfluoroundecyl methacrylate
CAS 41123-44-8
Molecular Formula C15H8F20O2
Formula Weight 600.2
MDL Number MFCD00042111
Synonym 1H,1H,11H-Eicosafluoroundecyl methacrylate; Methacrylic acid 1H,1H,11H-perfluoroundecyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1648906

 
alfa aesar™
L1648906
5g Each for $62.96

AAL1648914

 
alfa aesar™
L1648914
25g Each for $224.73
Description & Specifications

Specifications

Formula Weight 600.2
Chemical Name or Material 1H,1H,11H-Perfluoroundecyl methacrylate
Assay 95%
Molecular Formula C15H8F20O2
MDL Number MFCD00042111
CAS 41123-44-8
Synonym 1H,1H,11H-Eicosafluoroundecyl methacrylate; Methacrylic acid 1H,1H,11H-perfluoroundecyl ester
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Recommended Storage 2° to 8°C
Boiling Point 110°C (0.05mmHg)
Melting Point 36° to 38°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C