missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H,1H,11H-Perfluoroundecyl acrylate, tech. 85%, stab. with ca 50ppm 4-methoxyphenol

$51.97 - $175.80

Chemical Identifiers

CAS 4998-38-3
Molecular Formula C14H6F20O2
Molecular Weight (g/mol) 586.168
MDL Number MFCD00042112
InChI Key ASJLMYWCUSHRIW-UHFFFAOYSA-N
Synonym 1h,1h,11h-eicosafluoroundecyl acrylate, 1h,1h,11h-perfluoroundecyl acrylate, 1h,1h,11h-perfluoroundecyl acrylate, acrylic acid 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluoroundecyl ester
PubChem CID 2776419
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluoroundecyl prop-2-enoate
SMILES C=CC(=O)OCC(C(C(C(C(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1648806 View Documents Alfa Aesar™
L1648806
5g Each for $51.97
 
AAL1648814 View Documents Alfa Aesar™
L1648814
25g Each for $175.80
 
Specifications

Chemical Identifiers

4998-38-3
586.168
ASJLMYWCUSHRIW-UHFFFAOYSA-N
2776419
C=CC(=O)OCC(C(C(C(C(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
C14H6F20O2
MFCD00042112
1h,1h,11h-eicosafluoroundecyl acrylate, 1h,1h,11h-perfluoroundecyl acrylate, 1h,1h,11h-perfluoroundecyl acrylate, acrylic acid 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluoroundecyl ester
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluoroundecyl prop-2-enoate

Specifications

115°C (1mmHg)
5g
C14H6F20O2
1h,1h,11h-eicosafluoroundecyl acrylate, 1h,1h,11h-perfluoroundecyl acrylate, 1h,1h,11h-perfluoroundecyl acrylate, acrylic acid 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluoroundecyl ester
C=CC(=O)OCC(C(C(C(C(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
586.168
586.17
Technical
44°C to 53°C
4998-38-3
MFCD00042112
ASJLMYWCUSHRIW-UHFFFAOYSA-N
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluoroundecyl prop-2-enoate
2776419
85%
1H,1H,11H-Perfluoroundecyl acrylate, stabilized with ≈50ppm 4-methoxyphenol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C