missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1H,1H,10H,10H-Perfluoro-1,10-decanediol, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1682306
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1H,1H,10H,10H-Perfluoro-1,10-decanediol | |
132°C to 134°C | |
C10H6F16O2 | |
5 g | |
1h,1h,10h,10h-perfluorodecane-1,10-diol, 1h,1h,10h,10h-perfluoro-1,10-decanediol, 1h,1h,10h,10h-hexadecafluoro-1,10-decanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluoro-1,10-decanediol, 1,10-decanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluoro, hexadecafluorooctamethylenebismethanol, 1,10-decanediol,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluoro, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluoro-1,10-decandiol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecakis fluoranyl decane-1,10-diol | |
C(C(C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O | |
462.13 | |
462.13 |
754-96-1 | |
132°C (4 mmHg) | |
MFCD00192200 | |
1894671 | |
NSKCTPBWPZPFHW-UHFFFAOYSA-N | |
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluorodecane-1,10-diol | |
2733404 | |
96% |
Chemical Identifiers
754-96-1 | |
462.13 | |
NSKCTPBWPZPFHW-UHFFFAOYSA-N | |
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluorodecane-1,10-diol |
C10H6F16O2 | |
MFCD00192200 | |
2733404 | |
C(C(C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only