Please login to your online account to display your discounted pricing

Alfa Aesar™ 1H-1,2,4-Triazole-1-acetic acid, 97%

$104.58 - $290.81

Chemical Identifiers

Chemical Name or Material 1H-1,2,4-Triazole-1-acetic acid
CAS 28711-29-7
Molecular Formula C4H5N3O2
Formula Weight 127.1
MDL Number MFCD00036051
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3173203

  SDS
alfa aesar™
H3173203
1g Each for $290.81

AAH31732MD

  SDS
alfa aesar™
H31732MD
250mg Each for $104.58
Description & Specifications

Specifications

Formula Weight 127.1
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Molecular Formula C4H5N3O2
Health Hazard 1 Warning
Melting Point 201° to 205°C
MDL Number MFCD00036051
Assay 97%
Quantity 1g
Chemical Name or Material 1H-1,2,4-Triazole-1-acetic acid
CAS 28711-29-7

Applications
1H-1,2,4-Triazole-1-acetic acid is used to produce (1-hydroxy-1-phosphono-2-[1,2,4]triazol-1-yl-ethyl)-phosphonic acid at the temperature of 100 - 110°C. It will need reagent H3PO4, PCl3 and solvent chlorobenzene

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.