missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H-1,2,3-Triazole, TCI America™

Manufacturer:  TCI America T18805G

 View more versions of this product

Catalog No. T18805G



Specifications

Specifications

1H-1,2,3-Triazole
5g
C2H3N3
1h-1,2,3-triazole, 1,2,3-triazole, 2h-1,2,3-triazole, 1,2,3-1h-triazole, triazole, triazol, osotriazole, v-triazole, 1,2,3-triazol, 1,2,3 triazole
C1=NNN=C1
69.067
CHEBI:35565
Crystal-Lump at 20°C
White-Yellow
288-36-8
MFCD00014490
QWENRTYMTSOGBR-UHFFFAOYSA-N
2H-triazole
67516
69.07
≥98.0% (T)

Chemical Identifiers

288-36-8
69.067
QWENRTYMTSOGBR-UHFFFAOYSA-N
67516
2H-triazole
C2H3N3
MFCD00014490
1h-1,2,3-triazole, 1,2,3-triazole, 2h-1,2,3-triazole, 1,2,3-1h-triazole, triazole, triazol, osotriazole, v-triazole, 1,2,3-triazol, 1,2,3 triazole
CHEBI:35565
C1=NNN=C1
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6196

TSCA : No

Documents