missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
19-Maleimido-17-oxo-4,7,10,13-tetraoxa-16-azanonadecanoic Acid 95.0+%, TCI America™
$101.21 - $283.68
Chemical Identifiers
CAS | 1263045-16-4 |
---|---|
Molecular Formula | C18H28N2O9 |
Molecular Weight (g/mol) | 416.43 |
MDL Number | MFCD13184954 |
InChI Key | IAJVEYLYYHOZEY-UHFFFAOYSA-N |
Synonym | 1-Maleimido-3-oxo-7,10,13,16-tetraoxa-4-azanonadecan-19-oic Acid |
PubChem CID | 51340949 |
IUPAC Name | 1-[3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanamido]-3,6,9,12-tetraoxapentadecan-15-oic acid |
SMILES | OC(=O)CCOCCOCCOCCOCCNC(=O)CCN1C(=O)C=CC1=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
M266625MG
|
TCI America
M266625MG |
25 mg |
Each for $101.21
|
|
|||||
M2666100MG
|
TCI America
M2666100MG |
100 mg |
Each for $283.68
|
|
|||||
Chemical Identifiers
1263045-16-4 | |
416.43 | |
IAJVEYLYYHOZEY-UHFFFAOYSA-N | |
51340949 | |
OC(=O)CCOCCOCCOCCOCCNC(=O)CCN1C(=O)C=CC1=O |
C18H28N2O9 | |
MFCD13184954 | |
1-Maleimido-3-oxo-7,10,13,16-tetraoxa-4-azanonadecan-19-oic Acid | |
1-[3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanamido]-3,6,9,12-tetraoxapentadecan-15-oic acid |
Specifications
1263045-16-4 | |
MFCD13184954 | |
1-Maleimido-3-oxo-7,10,13,16-tetraoxa-4-azanonadecan-19-oic Acid | |
OC(=O)CCOCCOCCOCCOCCNC(=O)CCN1C(=O)C=CC1=O | |
416.43 | |
416.43 | |
Crystalline Powder |
C18H28N2O9 | |
25 mg | |
IAJVEYLYYHOZEY-UHFFFAOYSA-N | |
1-[3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanamido]-3,6,9,12-tetraoxapentadecan-15-oic acid | |
51340949 | |
≥95.0% (HPLC) | |
19-Maleimido-17-oxo-4,7,10,13-tetraoxa-16-azanonadecanoic Acid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator