missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
18-beta-Glycyrrhetinic acid, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 120150250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
18-β-Glycyrrhetinic acid | |
471-53-4 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD00003706,MFCD00066716 | |
15, 3644 | |
+ 165 | |
MPDGHEJMBKOTSU-YKLVYJNSSA-N | |
(2S,4aS,6aR,6aS,6bR,8aR,10S,12aS,14bR)-10-hydroxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,6a,7,8,8a,10,11,12,14b-dodecahydro-1H-picene-2-carboxylic acid | |
10114 | |
470.69 | |
2% max. (HPLC) |
98+% | |
98.0 | |
1% max. (105°C, 4 hrs) | |
98+% | |
C30H46O4 | |
25g | |
+165° (20°C c=1,CHC3,on dry ba) | |
enoxolone, glycyrrhetinic acid, uralenic acid, glycyrrhetic acid, 18-beta-glycyrrhetinic acid, glycyrrhetin, arthrodont, jintan, 18beta-glycyrrhetinic acid, rhetinic acid | |
CC1(C)[C@@H](O)CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O | |
470.69 | |
CHEBI:30853 | |
98+% |
Safety and Handling
EINECSNumber : 207-444-6
RTECSNumber : RK0180000
TSCA : TSCA
RUO â Research Use Only