missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
18-Crown-6, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 181560050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagentSpecifications
18-Crown-6 | |
36.0°C to 40.0°C | |
>110°C | |
98.5% min. (GC) | |
C12H24O6 | |
5 g | |
14,2602 | |
Solubility in water: soluble. Other solubilities: soluble in hydrocarbons,benzene,chloroform,methanol and trichlorotrifluoroethane | |
C1COCCOCCOCCOCCOCCO1 | |
264.32 | |
CHEBI:32397 | |
99% |
17455-13-9 | |
Colorless or White | |
Authentic | |
Glass bottle | |
MFCD00005113 | |
04,142; 05,152; 06,133; 07,76; 08,1280; 09,126; 12,403; 15,76; 17,290 | |
18-crown-6, 18-crown-6 ether, ethylene oxide cyclic hexamer, 18-crown-6-ether, unii-63j177nc5b, ccris 3587, 18-crown 6-ether, 1,4,7,10,13,16-hexanoxacyclooctadecane, 18-crown-6/1,4,7,10,13,16-hexaoxacyclooctadecane | |
XEZNGIUYQVAUSS-UHFFFAOYSA-N | |
1,4,7,10,13,16-hexaoxacyclooctadecane | |
28557 | |
264.32 | |
Crystalline Mass, Crystals or Liquid |
Chemical Identifiers
17455-13-9 | |
264.32 | |
XEZNGIUYQVAUSS-UHFFFAOYSA-N | |
28557 | |
1,4,7,10,13,16-hexaoxacyclooctadecane |
C12H24O6 | |
MFCD00005113 | |
18-crown-6, 18-crown-6 ether, ethylene oxide cyclic hexamer, 18-crown-6-ether, unii-63j177nc5b, ccris 3587, 18-crown 6-ether, 1,4,7,10,13,16-hexanoxacyclooctadecane, 18-crown-6/1,4,7,10,13,16-hexaoxacyclooctadecane | |
CHEBI:32397 | |
C1COCCOCCOCCOCCOCCO1 |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 241-473-5
RUO – Research Use Only