missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

17alpha-Hydroxyprogesterone, TCI America™

$110.00 - $330.00

Chemical Identifiers

CAS 68-96-2
Molecular Formula C21H30O3
Molecular Weight (g/mol) 330.468
MDL Number MFCD00003659
InChI Key DBPWSSGDRRHUNT-CEGNMAFCSA-N
Synonym 17alpha-Hydroxy-4-pregnene-3,20-dione
PubChem CID 6238
ChEBI CHEBI:17252
IUPAC Name (8R,9S,10R,13S,14S,17R)-17-acetyl-17-hydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
SMILES CC(=O)C1(CCC2C1(CCC3C2CCC4=CC(=O)CCC34C)C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H12505G View Documents TCI America
H12505G
5g Each for $110.00
 
H125025G View Documents TCI America
H125025G
25g Each for $330.00
 
Specifications

Chemical Identifiers

68-96-2
330.468
DBPWSSGDRRHUNT-CEGNMAFCSA-N
6238
(8R,9S,10R,13S,14S,17R)-17-acetyl-17-hydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
C21H30O3
MFCD00003659
17alpha-Hydroxy-4-pregnene-3,20-dione
CHEBI:17252
CC(=O)C1(CCC2C1(CCC3C2CCC4=CC(=O)CCC34C)C)O

Specifications

White
5g
C21H30O3
17alpha-Hydroxy-4-pregnene-3,20-dione
CC(=O)C1(CCC2C1(CCC3C2CCC4=CC(=O)CCC34C)C)O
330.468
CHEBI:17252
Crystal-Powder at 20°C
17alpha-Hydroxyprogesterone
220°C
68-96-2
MFCD00003659
DBPWSSGDRRHUNT-CEGNMAFCSA-N
(8R,9S,10R,13S,14S,17R)-17-acetyl-17-hydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
6238
330.47
≥98.0% (HPLC)
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1068

RTECSNumber : TU5060000

TSCA : No