Please login to your online account to display your discounted pricing

Alfa Aesar™ 17-Phenyl-trinor-prostaglandin E2, 98%

Chemical Identifiers

Chemical Name or Material 17-Phenyl-trinor-prostaglandin E2
CAS 38315-43-4
Formula Weight 386.49
MDL Number MFCD00135219
Synonym 17-Phenyl-trinor-PGE2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67126MCR

  SDS
alfa aesar™
J67126MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 386.49
MDL Number MFCD00135219
Chemical Name or Material 17-Phenyl-trinor-prostaglandin E2
Health Hazard 3 GHS P Statement
P281-P264-P301+P312-P308+P313-P405-P501a
Use personal protective equipment as required.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Quantity 1mg
Synonym 17-Phenyl-trinor-PGE2
RTECS Number MJ9681060
Color Colorless
Storage Note 1 -30° to -10°C
Physical Form Oil
Health Hazard 2 GHS H Statement
H360-H302
May damage fertility or the unborn child.
Harmful if swallowed.
Health Hazard 1 Danger
CAS 38315-43-4
Safety and Handling

Danger

GHS P Statement
P281-P264-P301+P312-P308+P313-P405-P501a
Use personal protective equipment as required.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H302
May damage fertility or the unborn child.
Harmful if swallowed.

-30° to -10°C