Please login to your online account to display your discounted pricing

Alfa Aesar™ 17-beta-Estradiol, BT-241

Chemical Identifiers

Chemical Name or Material 17-beta-Estradiol
CAS 50-28-2
Formula Weight 272.39
MDL Number MFCD00003693
Beilstein 1914275
Merck Index 14,3703
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ64137FPL

  SDS
alfa aesar™
J64137FPL
20mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 175° to 178°C
Formula Weight 272.39
MDL Number MFCD00003693
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 17-beta-Estradiol
Beilstein 1914275
RTECS Number KG2975000
CAS 50-28-2
Merck Index 14,3703
Health Hazard 2 GHS H Statement
H350-H360
May cause cancer.
May damage fertility or the unborn child.
Health Hazard 1 Danger
Physical Form Powder
Color White
Quantity 20mg
Safety and Handling

Danger

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H360
May cause cancer.
May damage fertility or the unborn child.