Please login to your online account to display your discounted pricing

Alfa Aesar™ 17-(Allylamino)-17-demethoxygeldanamycin, 99%


Quantity 200mg

Manufacturer: alfa aesar™  J66960EX3

 View more versions of this product

Catalog No. AAJ66960EX3

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 585.69
Melting Point 207° to 209°C
Molecular Formula C31H43N3O8
Recommended Storage −30° to −10°C
Chemical Name or Material 17-(Allylamino)-17-demethoxygeldanamycin
Physical Form Solid
MDL Number MFCD04973892
CAS 75747-14-7
Assay 99%
Synonym 17-AAG; 17-(Allylamino)geldanamycin
Sensitivity Light Sensitive
Storage Note 1 -30° to -10°C
RTECS Number LX8932000
Solubility Information Soluble in DMSO at 150mg/mL; soluble in ethanol at 5mg/mL; Very poorly soluble in water
Quantity 200mg

Specifications

Chemical Name or Material 17-(Allylamino)-17-demethoxygeldanamycin
CAS 75747-14-7
Molecular Formula C31H43N3O8
Formula Weight 585.69
MDL Number MFCD04973892
Synonym 17-AAG; 17-(Allylamino)geldanamycin

Solubility
Soluble in DMSO at 150 mg/mL; soluble in ethanol at 5 mg/mL. Very poorly soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Accessories

−30° to −10°C

-30° to -10°C

SDS & Safety Documents