Learn More
Chemglass Inc 16mL (4 Dram) Vial, 21mm OD x 70mm Height, 18-400 Thread Size
Supplier: Chemglass Inc CG490003
16mL (4 Dram) Vial, 21mm OD x 70mm Height, 18-400 Thread Size. Sample vials packaged in convenient LAB-PACS of 144. Made from Type 1 Borosilicate glass. Full height partitions minimize breakage. Solid PTFE Lined Caps.