Fisher Science Education™ 160-Series University/Laboratory Compound Microscopes
Designed for university and laboratory use. Fisherbrand™ 160-Series University/Laboratory Compound Microscope has good optical clarity and reliable mechanical engineering assure years of top performance.
Manufacturer:  Fisher Science Education™ 167ASC
 View more versions of this product
Catalog No. S23874
Description
Description
  • Remarkable optical clarity
  • Choice of objective lenses:  ASC, semi-plan, plan and phase
  • LED energy-efficient illumination 
  • Large, built-in, low-profile, rackless stage
  • Coaxial focusing
  • Magnification:  40X, 100X, 400X, and 1000X
Specifications
Specifications
Abbe 1.25 N.A
Super high contrast
LED
Microscope
40X, 100X, 400X, 1000X
Monocular
Lifetime limited warranty, excluding bulb, cord and fuse
Documents