Please login to your online account to display your discounted pricing

16-Hydroxyhexadecanoic acid, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  175900050

 View more versions of this product

Catalog No. AC175900050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 95° to 99°C
Color White
Quantity 5g
Chemical Name or Material 16-Hydroxyhexadecanoic acid
CAS 506-13-8
CAS Min % 97.5
CAS Max % 100.0
Physical Form White crystalline powder
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C16H32O3
Linear Formula HO(CH2)15CO2H
Formula Weight 272.42g/mol
MDL Number MFCD00002750
Beilstein 03, 362
Synonym Juniperic acid

Specifications

Chemical Name or Material 16-Hydroxyhexadecanoic acid
CAS 506-13-8
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C16H32O3
Linear Formula HO(CH2)15CO2H
Formula Weight 272.42g/mol
MDL Number MFCD00002750
Synonym Juniperic acid