Please login to your online account to display your discounted pricing

16-Hydroxyhexadecanoic acid, 98%, ACROS Organics™

$59.04 - $59.04

Chemical Identifiers

Chemical Name or Material 16-Hydroxyhexadecanoic acid
CAS 506-13-8
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C16H32O3
Linear Formula HO(CH2)15CO2H
Formula Weight 272.42g/mol
MDL Number MFCD00002750
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC175900050

  SDS
fisher scientific acros
175900050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC175900010

  SDS
fisher scientific acros
175900010
Glass bottle 1g Each for $59.04
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 95° to 99°C
Color White
Quantity 5g
Chemical Name or Material 16-Hydroxyhexadecanoic acid
CAS 506-13-8
CAS Min % 97.5
CAS Max % 100.0
Physical Form White crystalline powder
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C16H32O3
Linear Formula HO(CH2)15CO2H
Formula Weight 272.42g/mol
MDL Number MFCD00002750
Beilstein 03, 362
Synonym Juniperic acid