Please login to your online account to display your discounted pricing

16-Hydroxyhexadecanoic acid, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  175900010

 View more versions of this product

Catalog No. AC175900010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 272.42
Quantity 1g
Color White
Beilstein 03, 362
Melting Point 95° to 99°C
CAS Max % 100.0
Chemical Name or Material 16-Hydroxyhexadecanoic acid
Molecular Formula C16H32O3
CAS 506-13-8
Infrared Spectrum Authentic
Assay Percent Range 98%
MDL Number MFCD00002750
CAS Min % 97.5
Packaging Glass bottle
Linear Formula HO(CH2)15CO2H
Synonym Juniperic acid

Specifications

Chemical Name or Material 16-Hydroxyhexadecanoic acid
CAS 506-13-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C16H32O3
Linear Formula HO(CH2)15CO2H
Formula Weight 272.42
MDL Number MFCD00002750
Synonym Juniperic acid