Thermo Scientific™ Sliding Drawer Rack for Forma™ 88000 and TSU Series Freezers, holds 25 2 in. boxes
Maximize the storage space in your ultra-low temperature freezer with these sliding drawer racks, adjustable side access box racks and microplate racks.
Supplier: Thermo Scientific™ 920090DUPL
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for Forma™ 88000 and TSU Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
- With sliding drawer racks
- Adjustable side access box racks
- Standard/deepwell microplate racks