Thermo Scientific™ 150µL Polyspring Autosampler Insert, conical, pulled point, glass, 150μL Polyspring, Conical, Pulled point, Glass
Clear glass or polypropylene inserts for 8mm HPLC and GC autosampler vials are available in a variety of configurations with usable volumes from 125μL to 200μL.
Manufacturer:  Thermo Scientific™ C4012530
Catalog No. 35-653
Description
Description
These clear glass or polypropylene inserts for 8mm HPLC and GC autosampler vials are available in a variety of configurations with usable volumes ranging from 125-200μL.
  • Pulled point inserts are an economical choice for non-critical applications