Please login to your online account to display your discounted pricing

Alfa Aesar™ 15-Deoxy-delta12,14-prostaglandin J2, 97%


Quantity 1mg

Manufacturer: alfa aesar™  J67427MCR

 View more versions of this product

Catalog No. AAJ67427MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 316.44
Chemical Name or Material 15-Deoxy-delta12,14-prostaglandin J2
Physical Form Oil
Assay 97%
MDL Number MFCD00065806
Synonym 15-Deoxy-PGJ2
Storage Note 1 -30° to -10°C
Quantity 1mg
CAS 87893-55-8

Specifications

Chemical Name or Material 15-Deoxy-delta12,14-prostaglandin J2
CAS 87893-55-8
Formula Weight 316.44
MDL Number MFCD00065806
Synonym 15-Deoxy-PGJ2
Accessories

-30° to -10°C