missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-4-Hydroxystilbene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 121930050

 View more versions of this product

Catalog No. AC121930050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

trans-4-Hydroxystilbene
6554-98-9
96.5% min. (HPLC)
C14H12O
MFCD00002386
06,693
Solubility in water: insoluble. Other solubilities: soluble in acetone,chloroform,benzene,,soluble in acetic acid
C1=CC=C(C=C1)C=CC2=CC=C(C=C2)O
196.25
CHEBI:35101
98%
98%
185°C to 189°C
Glass Bottle
C6H5CH=CHC6H4OH
5 g
trans-4-hydroxystilbene, 4-hydroxystilbene, 4-styrylphenol, 4-stilbenol, e-4-stilbenol, 4-2-phenylethenyl phenol, 4-2-phenylvinyl phenol, 4-e-2-phenylethenyl phenol, e-4-hydroxystilbene, stilben-4-ol
QVLMUEOXQBUPAH-VOTSOKGWSA-N
4-[(E)-2-phenylethenyl]phenol
5284650
196.25

Chemical Identifiers

6554-98-9
196.25
QVLMUEOXQBUPAH-VOTSOKGWSA-N
5284650
4-[(E)-2-phenylethenyl]phenol
C14H12O
MFCD00002386
trans-4-hydroxystilbene, 4-hydroxystilbene, 4-styrylphenol, 4-stilbenol, e-4-stilbenol, 4-2-phenylethenyl phenol, 4-2-phenylvinyl phenol, 4-e-2-phenylethenyl phenol, e-4-hydroxystilbene, stilben-4-ol
CHEBI:35101
C1=CC=C(C=C1)C=CC2=CC=C(C=C2)O
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 229-483-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only