Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ 14.3.3 pan Ab-4, Mouse Monoclonal Antibody

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ 14.3.3 pan Ab-4, Mouse Monoclonal Antibody.

$271.27 - $1,369.49

Specifications

Antigen 14.3.3 pan Ab-4
Clone CG15
Applications Immunoprecipitation
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1504P

 
thermo scientific™ lab vision™
MS1504P
200μg/mL 1mL Each for $1,256.61

MS1504P0

 
thermo scientific™ lab vision™
MS1504P0
200μg/mL 100μL Each for $271.27

MS1504P1

 
thermo scientific™ lab vision™
MS1504P1
200μg/mL 500μL Each for $760.47

MS1504P1ABX

 
thermo scientific™ lab vision™
MS1504P1ABX
1mg/mL 100μL Each for $853.59

MS1504PABX

 
thermo scientific™ lab vision™
MS1504PABX
1mg/mL 200μL Each for $1,369.49
Description & Specifications

Specifications

Antigen 14.3.3 pan Ab-4
Clone CG15
Applications Immunoprecipitation
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Signal Transduction
Type Primary
Immunogen 14-3-3 gamma recombinant human protein
Isotype IgG2a kappa

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ 14.3.3 pan Ab-4, Mouse Monoclonal Antibody.

14.3.3 proteins are a family of small, widely expressed, highly conserved cytosolic proteins. 14.3.3 proteins bind to and influence the activities of a diverse group of molecules involved in signal transduction, cell cycle regulation and apoptosis, including Raf, PKC, Bad, Cbl, and c-Bcr. Interactions between 14.3.3 and target proteins are strongly influenced by the phosphorylation state of 14.3.3 and the target protein.

Host Species: Mouse

Clone: CG15

Isotype: IgG1/k

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: 14.3.3γ recombinant human protein

Molecular Weight: 26-33kDa

Positive Control: HeLa cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO