Please login to your online account to display your discounted pricing

Thermo Scientific™ 13mm Certified Clear Screw Thread Kits

These 13mm vials are 4mL, 15x45mm vials with 13-425 thread finish. They are manufactured from high-quality glass and fit a variety of autosamplers.

$187.78 - $187.78

Specifications

Closure Material Bonded PTFE/Silicone
Description Certified 4mL Clear Glass Screw-top Vial Kit, 13-425 Screw Thread
Closure Type Screw Thread
Color Clear
Material Glass
View More Specs
Products
Catalog Number Mfr. No. Closure Type Type Price Quantity    

03-377-352

 
thermo scientific™
CERT401583
Screw Thread Unassembled Vial Kit Pack of 100 Each for $187.78
Description & Specifications

Specifications

Closure Material Bonded PTFE/Silicone
Description Certified 4mL Clear Glass Screw-top Vial Kit, 13-425 Screw Thread
Closure Type Screw Thread
Color Clear
Material Glass
No. per Pack 100
Patched No
Type Unassembled Vial Kit
Item Description Convenience Kit, Certified 13-425 Screw Vial, 4mL
Septa PTFE/Silicone
Vial Closure Size 13-425mm
Vial Volume (Metric) 4mL

These 13mm vials are 4mL, 15x45mm vials with 13-425 thread finish. Manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) glass and feature a 13-425 thread finish, they are available in Thermo Scientific™ Premiere™, National™ and Chromacol™ brands.

Choose the right vial for your needs

  • Get maximum sample extraction without need for separate inserts by using Microsampling and High Recovery Vials
  • Identify sample and volume using the graduated, write-on patch provided on certain vials
  • Obtain maximum recovery of critical polar, labile or OH-interacting compounds using silanized (deactivated) glass (optional)

Choose the best insert for your sample needs

  • Polyspring inserts are self-aligning and provide a cushion against needle contact
  • Precision point insert minimizes residual sample loss

Autosampler compatibility*

  • Jasco™
  • PerkinElmer™
  • Shimadzu™
  • Varian™
  • Waters™

*See the Thermo Scientific Columns and Consumables Catalog for autosampler compatibility