Please login to your online account to display your discounted pricing

Alfa Aesar™ 13(S)-Hydroxyoctadeca-9Z,11E-dienoic acid, 98%, 90-100 μg/mL in ethanol

Chemical Identifiers

Chemical Name or Material 13(S)-Hydroxyoctadeca-9Z,11E-dienoic acid
CAS 29623-28-7
Name Note 90 to 100μg/mL in ethanol
Formula Weight 296.45
MDL Number MFCD00065833
UN Number UN1170
Synonym (S)-13-HODE
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67148EXD

 
alfa aesar™
J67148EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67148EXG

 
alfa aesar™
J67148EXG
100μg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 296.45
Chemical Name or Material 13(S)-Hydroxyoctadeca-9Z,11E-dienoic acid
MDL Number MFCD00065833
CAS 29623-28-7
Health Hazard 3 GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Name Note 90 to 100μg/mL in ethanol
Physical Form Liquid
Assay 98%
UN Number UN1170
Synonym (S)-13-HODE
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H371
Highly flammable liquid and vapor.
May cause damage to organs.
DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: ETHANOL SOLUTIONS
Storage Note 1 -30° to -10°C
Quantity 0.5mg
Safety and Handling

Danger

GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H371
Highly flammable liquid and vapor.
May cause damage to organs.

-30° to -10°C