Please login to your online account to display your discounted pricing

Alfa Aesar™ 13(S)-Hydroxyoctadeca-9Z,11E-dienoic acid, 98%, 90-100 μg/mL in ethanol


Quantity 100μg

Manufacturer: alfa aesar™  J67148EXG

 View more versions of this product

Catalog No. AAJ67148EXG

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 296.45
CAS 29623-28-7
MDL Number MFCD00065833
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H371
Highly flammable liquid and vapor.
May cause damage to organs.
Health Hazard 3 GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: ETHANOL SOLUTIONS
Chemical Name or Material 13(S)-Hydroxyoctadeca-9Z,11E-dienoic acid
Name Note 90 to 100μg/mL in ethanol
Physical Form Liquid
Assay 98%
UN Number UN1170
Synonym (S)-13-HODE
Storage Note 1 -30° to -10°C
Quantity 100μg

Specifications

Chemical Name or Material 13(S)-Hydroxyoctadeca-9Z,11E-dienoic acid
CAS 29623-28-7
Name Note 90 to 100μg/mL in ethanol
Formula Weight 296.45
MDL Number MFCD00065833
UN Number UN1170
Synonym (S)-13-HODE
Accessories

Danger

GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H371
Highly flammable liquid and vapor.
May cause damage to organs.

-30° to -10°C

SDS & Safety Documents