missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

13-cis-Retinoic Acid 97.0+%, TCI America™

Manufacturer:  Tci America R0088100MG

 View more versions of this product

Catalog No. R0088100MG



Specifications

Specifications

13-cis-Retinoic Acid
175°C
4759-48-2
MFCD00079542
SHGAZHPCJJPHSC-RVDNBNLISA-N
(2Z,4E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
131954766
Crystal-Powder at 20°C
Undesignated
100mg
C20H28O2
(2Z,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2,4,6,8-nonatetraenoic Acid, Isotretinoin
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)O)C)C
300.442
300.44
≥97.0% (HPLC,T)

Chemical Identifiers

4759-48-2
300.442
SHGAZHPCJJPHSC-RVDNBNLISA-N
131954766
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)O)C)C
C20H28O2
MFCD00079542
(2Z,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2,4,6,8-nonatetraenoic Acid, Isotretinoin
(2Z,4E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
Safety and Handling

Safety and Handling

RTECSNumber : VH6440000

TSCA : Yes

Recommended Storage : Freezer

Documents
Promotions

Promotions