Please login to your online account to display your discounted pricing

Alfa Aesar™ (±)13-Azaprostanoic acid, 98%

Chemical Identifiers

Chemical Name or Material (±)13-Azaprostanoic acid
CAS 71629-07-7
Formula Weight 311.51
MDL Number MFCD00077317
Synonym 13-APA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67164MCR

  SDS
alfa aesar™
J67164MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 311.51
Chemical Name or Material (±)13-Azaprostanoic acid
CAS 71629-07-7
MDL Number MFCD00077317
Color White
Assay 98%
Physical Form Solid
Synonym 13-APA
Storage Note 1 -30° to -10°C
Quantity 1mg
Safety and Handling

-30° to -10°C