missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(±)13-Azaprostanoic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J67164MCR
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(±)13-Azaprostanoic acid | |
White | |
MFCD00077317 | |
13-azaprostanoic acid, 7-1s,2s-2-heptylamino cyclopentyl heptanoic acid, 13-azaprostan-1-oic acid, chembl9811, bml1-g10, +/-13-azaprostanoic acid, cyclopentaneheptanoicacid, 2-heptylamino-, 1s,2s, cyclopentaneheptanoic acid, 2-heptylamino-, 1s-trans | |
CCCCCCCNC1CCCC1CCCCCCC(=O)O | |
311.51 | |
311.51 | |
Solid |
71629-07-7 | |
C19H37NO2 | |
1 mg | |
RZZNGDIYQSCFGK-ROUUACIJSA-N | |
7-[(1S,2S)-2-(heptylamino)cyclopentyl]heptanoic acid | |
123640 | |
98% |
Chemical Identifiers
71629-07-7 | |
311.51 | |
RZZNGDIYQSCFGK-ROUUACIJSA-N | |
123640 | |
CCCCCCCNC1CCCC1CCCCCCC(=O)O |
C19H37NO2 | |
MFCD00077317 | |
13-azaprostanoic acid, 7-1s,2s-2-heptylamino cyclopentyl heptanoic acid, 13-azaprostan-1-oic acid, chembl9811, bml1-g10, +/-13-azaprostanoic acid, cyclopentaneheptanoicacid, 2-heptylamino-, 1s,2s, cyclopentaneheptanoic acid, 2-heptylamino-, 1s-trans | |
7-[(1S,2S)-2-(heptylamino)cyclopentyl]heptanoic acid |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only