Isopropyl Alcohol, 70% v/v, Solvent Grade, Ricca Chemical Company™
$258.91 - $7,789.97
Chemical Identifiers
Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
R42100704A View Documents Ricca Chemical Company
R42100704A
4 L Poly Bottle Each for $258.91
R421007020F View Documents Ricca Chemical Company
R421007020F
20 L Cubitainer™ Each for $864.66
R421007055E View Documents Ricca Chemical Company
R421007055E
55 Gallons Drum Each for $7,789.97
Specifications
Chemical Identifiers
0.0
icon-pdf.png
Specifications
Colorless liquid
Poly Bottle
Colorless
Isopropyl Alcohol
68 to 72% v/v
At 20°C: 0.874-0.882
4 L