missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 12-Tungstophosphate hydrate, Reagent Grade

Manufacturer:  Alfa Aesar™ 1182730

 View more versions of this product

Catalog No. AA1182730



Specifications

Specifications

12-Tungstophosphate hydrate
H5O41PW12
UN3260
AVFBYUADVDVJQL-UHFFFAOYSA-N
phosphoric acid;trioxotungsten;hydrate
16212977
Powder
Moisture & Light Sensitive
95°C
12501-23-4
MFCD00011342
12-tungstophosphoric acid hydrate
O.OP(=O)(O)O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O
2898.053
2880.20 (Anhydrous)
Reagent
White through Yellow
250g

Chemical Identifiers

12501-23-4
2898.053
AVFBYUADVDVJQL-UHFFFAOYSA-N
16212977
O.OP(=O)(O)O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O
H5O41PW12
MFCD00011342
12-tungstophosphoric acid hydrate
phosphoric acid;trioxotungsten;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Documents