Please login to your online account to display your discounted pricing

Alfa Aesar™ 12-Tricosanone, 97%

Antiblocking agent

$233.55 - $233.55

Chemical Identifiers

Chemical Name or Material 12-Tricosanone
CAS 540-09-0
Formula Weight 338.62
MDL Number MFCD00026562
Synonym Diundecyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4144422

  SDS
alfa aesar™
4144422
100g Each for $233.55
Description & Specifications

Specifications

Formula Weight 338.62
Chemical Name or Material 12-Tricosanone
TSCA TSCA
Physical Form Powder
Assay 97%
MDL Number MFCD00026562
CAS 540-09-0
Synonym Diundecyl ketone
Melting Point 66° to 68°C
RTECS Number YD0805000
Quantity 100g