Learn More
12-Molybdophosphoric acid hydrate, ACS, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 05616606
Description
12-Molybdophosphoric acid hydrate is used as a precursor to hybrid organic-inorganic based molecular batteries with application as cation-insertion electrode. It finds application as an analytical reagent in thin layer chromatography for staining phenolics, hydrocarbon waxes, alkaloids and steroids. It acts as a reactant for the preparation of catalysts for the nitration of aromatic compounds, highly dispersed MoOx/CNTs (carbon nanotubes) as a support for Pt catalysts and anchored Wilkinson catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
12-Molybdophosphoric acid hydrate | |
78°C to 90°C | |
3.15 | |
H3Mo12O40P | |
5 g | |
Phosphomolybdic acid | |
FEFSFHGZSNHJEL-UHFFFAOYSA-N | |
phosphoric acid dodecamolybdenum hexatriacontaoxidandiide | |
1825.25 (Anhydrous) | |
Granules |
51429-74-4 | |
Yellow | |
Odorless | |
MFCD00149913 | |
Moisture sensitive | |
Soluble in water,ethanol and ether. | |
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].OP(O)(O)=O | |
1825.40 | |
ACS |
Chemical Identifiers
51429-74-4 | |
1825.40 | |
FEFSFHGZSNHJEL-UHFFFAOYSA-N | |
phosphoric acid dodecamolybdenum hexatriacontaoxidandiide |
H3Mo12O40P | |
MFCD00149913 | |
Phosphomolybdic acid | |
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].OP(O)(O)=O |
Safety and Handling
P210-P220-P221-P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
H272-H290-H314-H335-H373
DOTInformation : DOT Class: 8; Packaging Group: II
EINECSNumber : 234-713-5
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only