Please login to your online account to display your discounted pricing

12-Molybdophosphoric acid hydrate, ACS, Alfa Aesar™

$88.59 - $88.59

Chemical Identifiers

Chemical Name or Material 12-Molybdophosphoric acid hydrate
CAS 51429-74-4
Purity Grade ACS Grade
Molecular Formula H3PO4·12MoO3·xH2O
Formula Weight 1825.25 (Anhydrous)
MDL Number MFCD00149913
UN Number UN3085
Merck Index 14,7342
Synonym Phosphomolybdic acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA5616614

  SDS
alfa aesar
5616614
Each for $88.59
Description & Specifications

Specifications

Formula Weight 1825.25 (Anhydrous)
Purity Grade ACS Grade
Melting Point 78° to 90°C
Quantity 25g
Sensitivity Moisture Sensitive
CAS 51429-74-4
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P221-P280-P305+P351+P338-P308+P313-P501a;
Take any precaution to avoid mixing with combustibles.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00149913
Chemical Name or Material 12-Molybdophosphoric acid hydrate
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.
Synonym Phosphomolybdic acid
Merck Index 14,7342
Health Hazard 2 GHS H Statement
H272-H314-H318;
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Molecular Formula H3PO4·12MoO3·xH2O
Density 3.15
TSCA TSCA
Color Yellow
UN Number UN3085
Physical Form Granular
Safety and Handling

Danger

GHS P Statement
P221-P280-P305+P351+P338-P308+P313-P501a;
Take any precaution to avoid mixing with combustibles.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H314-H318;
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.