Please login to your online account to display your discounted pricing

Alfa Aesar™ 12-Methoxydodecanoic acid, 98%


Quantity 10mg

Manufacturer: alfa aesar™  J67495MA

 View more versions of this product

Catalog No. AAJ67495MA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 230.35
Melting Point 45° to 46°C
Chemical Name or Material 12-Methoxydodecanoic acid
MDL Number MFCD00899847
CAS 92169-28-3
Physical Form Solid
Assay 98%
Color White
Quantity 10mg

Specifications

Chemical Name or Material 12-Methoxydodecanoic acid
CAS 92169-28-3
Formula Weight 230.35
MDL Number MFCD00899847