Please login to your online account to display your discounted pricing

Alfa Aesar™ 12-Chloro-5-dodecyne, 98%

$69.92 - $69.92

Chemical Identifiers

Chemical Name or Material 12-Chloro-5-dodecyne
CAS 42513-36-0
Molecular Formula Cl(CH2)6C≡C(CH2)3CH3
Formula Weight 198.74
MDL Number MFCD00464356
Synonym n-Butyl 6-chlorohexyl acetylene; 1-Chloro-7-dodecyne
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2073006

  SDS
alfa aesar™
B2073006
5g Each for $69.92
Description & Specifications

Specifications

Formula Weight 198.74
Assay 98%
Quantity 5g
CAS 42513-36-0
TSCA TSCA
MDL Number MFCD00464356
Chemical Name or Material 12-Chloro-5-dodecyne
Synonym n-Butyl 6-chlorohexyl acetylene; 1-Chloro-7-dodecyne
Molecular Formula Cl(CH2)6C≡C(CH2)3CH3